Would you like to log in again?
One-Fit N95 Health Care Particulate Respirator, HONEYWELL
Supplier: HONEYWELL SAFETY PRODUCTS USA INC.
14110452 14110444 14110445
The ONE-Fit disposable N95 is a universal, single-size respirator designed to fit most face sizes and shapes.
  • Optional exhalation valve expels warm air
  • Strong and rigid outer cover prevents the mask from collapsing in environments with high humidity or fog
  • The two welded straps for the head keep the respirator comfortably on the face of the user
  • Risks: Air particles, contamination
Certifications: Meets NIOSH and CDC standards of N95 protection against pathogens transported by aure and M.tuberculosis
Packaging: Molded cup (20 masks/box); Molded cup with valve (10 masks/box); Smooth fold (20 masks/box)
ORDER
Expand All / Collapse All
Description Supplier No. VWR Catalog Number Unit Price Quantity
NBW95, Particulate Respirator With Molded Cup 14110444 89089-188 Case of 200 Retrieving
Pack of 20 Retrieving
W1400, flat folding particulate respirator 14110452 97039-398 Box of 20 Retrieving
NBW95V exhalation valve, particulate respirator with molded cup 14110445 89089-190 Case of 100 Retrieving
Pack of 10 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty