Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BRAND® TipBox/Tip Refill Environmentally-Friendy Filtered Pipette Tip System, BrandTech® Scientific

Supplier: BRANDTECH SCIENTIFIC INC.
Brand®
732710 732612 732602 732712 732614 732702 732604 732714 732704 732606 732706 732608 732708 732610
Low waste nonsterile tips fit BRAND® Transferpette® S and other leading pipettes.

  • Aerosol barrier to protect samples and pipettes
  • Non-sealing tips are free from contaminating additives
  • Environmentally-friendly, recyclable refillable boxes and inserts
  • Autoclavable
  • Oleamide- and diHEMDA-free

BRAND® filtered pipette tips are free from sealing additives that can contaminate samples and allow easy sample recovery

TipBoxes and TipRefills make up an environmentally friendly reloading system. TipRefills come with a transfer aid that enables simple, contamination-free transfer. TipBox lid comes installed as hinged, but can be removed and rotated 180⁰ to act as a lift off lid. Extra-rigid carrier plate is rimless to facilitate partial loading of multi-channel pipettes. Use of refills reduces waste by over 20%.

Tips are preminum quality, made from oleamide and diHEMDA-free resin, in one of the world's largest life science plastics cleanrooms. They are designed to fit most major brands. The nano-cap™ tips fit BRAND® 0.1 – 1 µL sub-micro Transferpette®, as well as most pipettes using ultra-micro or "crystal" tips. The 0.5 – 10 µL and 1 – 20 µL tips also fit pipettes using ultra-micro or "crystal" tips. The 2 – 20 µL, 5 – 100 µL, and 5 – 200 µL tips fit pipettes using standard 200 – 300 µL "Yellow" tips. The 50 – 1000 µL tips fit pipettes using standard 1000 µL "Blue" tips.

Filters made of pure polyethylene mean tips can be autoclaved at 121 °C for 20 minutes per DIN EN 285, without compromise. Tips are CE marked according to IVD-Directive 98/79 EC.

Ordering information: TipBoxes and carrier plates are made of polypropylene. TipRefill packaging is made of PET and medical paper. All components are recyclable.



 

ORDER

Expand All / Collapse All
Description Volume Packaging Packed Supplier No. VWR Catalog Number Unit Price Quantity
nano-cap™ Tips 0.1 – 1 µL TipBox 5 Boxes/480 Tips 732702 75831-148 Pack of 480 Retrieving
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
nano-cap™ Tips 0.1 – 1 µL TipRefill 10 Inserts/960 Tips 732602 75831-146 Pack of 960 Retrieving
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
Ultra-micro Tips 0.5 – 10 µL TipBox 5 Boxes/480 Tips 732704 75831-152 Pack of 480 Retrieving
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
Ultra-micro Tips 0.5 – 10 µL TipRefill 10 Inserts/960 Tips 732604 75831-150 Pack of 960 Retrieving
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
Ultra-micro Tips 1 – 20 µL TipBox 5 Boxes/480 Tips 732706 75831-156 Pack of 480 Retrieving
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
Ultra-micro Tips 1 – 20 µL TipRefill 10 Inserts/960 Tips 732606 75831-154 Pack of 690 Retrieving
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
"Yellow" Tips 2 – 20 µL TipBox 5 Boxes/480 Tips 732708 75831-160 Pack of 480 Retrieving
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
"Yellow" Tips 2 – 20 µL TipRefill 10 Inserts/960 Tips 732608 75831-158 Pack of 960 Retrieving
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
"Yellow" Tips 5 – 100 µL TipBox 5 Boxes/480 Tips 732710 75831-140 Pack of 480 Retrieving
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
"Yellow" Tips 5 – 100 µL TipRefill 10 Inserts/960 Tips 732610 75831-138 Pack of 960 Retrieving
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
"Yellow" Tips 5 – 200 µL TipBox 5 Boxes/480 Tips 732712 75831-144 Pack of 480 Retrieving
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
"Yellow" Tips 5 – 200 µL TipRefill 10 Inserts/960 Tips 732612 75831-142 Pack of 960 Retrieving
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
"Blue" Tips 50 – 1000 µL TipBox 5 Boxes/480 Tips 732714 75830-338 Pack of 480 Retrieving
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
"Blue" Tips 50 – 1000 µL TipRefill 10 Inserts/960 Tips 732614 75830-336 Pack of 960 Retrieving
Product and/or packaging is recyclable or reduces waste
Product and/or packaging is safer to human and environmental health
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Environmentally Preferable Attribute Product and/or packaging is recyclable or reduces waste, Product and/or packaging is safer to human and environmental health
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping