Would you like to log in again?
PolyScience® WhisperCool™ Refrigerated Chiller, Perkin Elmer
Supplier: PERKIN ELMER
N0772046 N0772045
10138-126EA 6522.41 USD InStock
10138-126 10138-124
PolyScience® WhisperCool™ Refrigerated Chiller, Perkin Elmer
Circulating Baths Closed Circulator Systems Cooling Circulators
The PolyScience® WhisperCool™ Refrigerated Chiller is designed to deliver quiet and reliable performance over a broad range of operating temperatures and conditions.
  • Dependable and energy efficient
  • Highly intuitive user interface
  • Operating temperature range –10 to +40 °C
  • Built-in process and equipment protection
  • Built-in temperature, pressure, and flow rate alarms
  • Operation is 50% quieter than comparable models (62 dB)
This chiller is designed for use on both ICP-OES and ICP-MS instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty