Iodine-125, MP Biomedicals
016303720 016303725 016303710 016303701 016303705 016303702
76169-538EA 1185.51 USD
76169-538 76169-536 76169-530 76169-540 76169-534 76169-532
Iodine-125, MP Biomedicals
Reliable performance and consistent quality.
  • NaI in 0.01 N NaOH, pH 8-11
  • Volume 0.1 ml solution unless otherwise specified.
MP Biomedicals' production processes and quality control systems ensure that our radiochemical products perform consistently and reliably.