Deoxycytidine-5'-Triphosphate, [α-³²P] , MP Biomedicals
0133004.2 0133004.5 013300401
76169-506EA 812.32 USD
76169-506 76169-504 76169-502
Deoxycytidine-5'-Triphosphate, [α-³²P] , MP Biomedicals
Reliable performance and consistent quality.
  • (dCTP α-32P)
  • Tetra-triethylammonium Salt
  • Sp. Act. 25 Ci/mmol; 925 GBq/mmol
  • 5 mM Tris HCl, pH 7.4-7.5
  • 10 mCi/ml; 370 MBq/ml
Radiolabeled Nucleotide with a molecular weight of 871.9. MP Biomedicals' production processes and quality control systems ensure that our radiochemical products perform consistently and reliably.