Soybean oil, orange oil
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Sojae Oleum
, Glycine Soja (Soybean) Oil
Soybean oil (SBO) is a vegetable oil widely used as lubricant. It may be transesterified to biodiesel using CaO as a solid base catalyst. It may also be transesterified to biodiesel using methanol and ethanol.
Soybean Oil may be used as a crude source for triglycerides and other lipids. Soybean oil is a potential raw material in oxygenated fatty acid production.
Soybean oil increases left ventricular contractility associated myosin ATPase and sarcoplasmic reticulum calcium pump (SERCA2a) in isolated hearts from rats. SBO is shown to reduce both total and non-HDL serum cholesterol and triglycerides induced by atherogenic diet in rats.
Store at Room Temperature (15-30 °C).
- Presentation: Orange Oil
- Refractive Index: n20/D 1.4743 (lit.)
- Miscible with alcohol, ether, petroleum ether; insoluble in water.
Melting Pt:
15…20 °F (lit.) Density: 0.917 g/mL (25 °C) . 0.92 g/mL (25 °C) (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00132356 CAS Number: 8001-22-7 |
Specification Test Results
Identity Test | Passes |
FTIR | Conforms to Standard |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...