3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS)

Synonyms: CHAPS

17-1314-01 17-1314-01
95017-118EA 107.06 USD
95017-118 CA95017-118L
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS)
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)

A nonionic or zwitterionic detergent is included in the sample solution to ensure complete sample solubilization and to prevent aggregation through hydrophobic interactions.


CHAPS and Triton X-100 are detergents commonly used in IEF.

Formula: C₃₂H₅₈N₂O₇S
MW: 614.89 g/mol
Melting Pt: 116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00081083
CAS Number: 75621-03-3
Merck Index: 12,02085

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR