Dithiothreitol (DTT, Cleland's reagent) for electrophoresis
|
Warning
|
Synonyms:
DL-1,4-Dithiothreitol
, DTT
, cis-2,3-Dihydroxy-1,4-butanedithiol
, (R*,R*)-1,4-Dimercapto-2,3-butandiol
17-1318-02
17-1318-01
17-1318-01
17-1318-02
95017-128EA
138.21
USD
CA95017-126L
95017-128
CA95017-128L
95017-126
Dithiothreitol (DTT, Cleland's reagent) for electrophoresis
Dithiothreitol (DTT, Cleland's reagent)
Reducing agents are frequently included in the sample solution to break any disulfide bonds present and to maintain all proteins in their fully reduced state. The most commonly used reductant is dithiothreitol (DTT).
Formula:
C₄H₁₀O₂S₂ MW: 154.25 g/mol Boiling Pt: ∼125 °C (1013 hPa) Melting Pt: 42…44 °C Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00004877 CAS Number: 3483-12-3 EINECS: 222-468-7 |
Specification Test Results
Application | IEF, SDS-PAGE |
Color | White |
Estimated shelf life from manufacture date | 4 years |
Melting/Freezing Point | 38-43°C |
Molecular Weight (Mr) | 154.24 |
pH Range | 0-5.1 |
Physical State | Solid |
Solubility | Easily soluble in cold water and hot water |
Storage conditions | 2-8°C |
Wetted Part | No |
DIGE approved | Y |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...