ICNA02194813

0219481325 0219481310 0219481380
76177-934EA 243.77 USD
76177-934 76177-932 76177-936
ICNA02194813
Storage: Store at -20 °C. Store Desiccated. Protect from light.
5-Bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt is β-glucuronidase substrate which forms an intense blue precipitate upon enzymatic action.
5-Bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt is used for the detection of the GUS gene in bacterial colonies and in histochemical applications.

  • Molecular Biology Reagent Grade
  • Presentation: White Powder
  • Note: Protect from light and humidity
  • Soluble in DMF and water (10 mg/mL - clear, practically colorless solution).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR