Ammonium persulfate (APS) ≥98%, white crystalline powder for molecular biology
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Danger
|
Synonyms:
di-Ammonium peroxodisulfate
, Ammonium persulfate
, Ammonium persulphate
, Diammonium peroxodisulfate
Storage: Store at Room Temperature (15-30 °C), dessicate.
Ammonium persulfate is a strong oxidizing agent. It is a radical initiator.
Ammonium persulfate is a polymerization catalyst used with TEMED for polyacrylamide gel formation. APS has also been utilized to study protein-protein interactions via photoinitiated crosslinking chemistry. Other applications of APS include its use as a reducer and retarder in photography, the manufacture of aniline dyes, electroplating, the decolorizing and deodorizing of oils.
- Molecular Biology Reagent Grade
- Presentation: White Crystalline Powder
- Vapor Density: 7.9 (vs air) (lit.)
- Soluble in water.
Formula:
(NH₄)₂S₂O₈ MW: 228.2 g/mol Melting Pt: 120 °C (dec., lit.) Density: 950…1050 kg/m³ (bulk density) Storage Temperature: Ambient |
MDL Number:
MFCD00003390 CAS Number: 7727-54-0 UN: 1444 ADR: 5.1,III |
Specification Test Results
Identity Test | Passes |
Purity | ≥98% |
FTIR | Conforms to Standard |
pH (1% aq soln) | Report Result |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...