Air-Supply Hose, MSA
Supplier: MSA
454-481080
300081-080EA 703.4 USD
300081-080
Air-Supply Hose, MSA
Tubing Hose
  • Chemical-resistant, black neoprene
  • Stainless steel couplings
  • Operating temperature range: –25 to 212 °F
Certifications: NIOSH Approved.