Weiler® Miniature Stem Mounted Wheel Brush, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-26014 804-26016 804-26021 804-26000 804-26296 804-26015 804-26144 804-26005
300049-604EA 1336.32 USD
300049-604 300050-050 300049-602 300049-610 300091-888 300050-052 300050-074 300050-054
Weiler® Miniature Stem Mounted Wheel Brush, ORS Nasco
Brushes
Miniature stem-mounted wheel brushes are a great choice for cleaning and deburring with a flexible shaft or straight grinder.
Brushes can be used with high-speed electric rotary tools, and air-powered pencil die grinders.
Use for buffing and polishing small parts. Also good for decorative finishing and edge blending.