ExoStd™ HCT116 Fluorescent Exosome Standard, BioVision Inc.
M1083-100
76177-580EA 702.98 USD
76177-580
ExoStd™ HCT116 Fluorescent Exosome Standard, BioVision Inc.
Green fluorescent Exosomes from HCT116 cell line.
Highly purified Exosomes available from HCT116 Cell line (Human Colon Carcinoma).