Hydrochloric acid fuming 37%, tested according to Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Muriatic acid

84436-1L
BJ84436-1LEA 93.31 USD
BJ84436-1L
Hydrochloric acid fuming 37%, tested according to Ph. Eur., Fluka™
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 42 °C (1.013 hPa)
Melting Pt: -35 °C
Density: 1.190 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Vapor Pressure 3.23 psi ( 21.1 °C)
Identity A complying
Identity B complying
Assay 35.0 - 39.0 %
Appearance of the solution complying
Non-volatile matter Max. 0.01 %
Sulfate (SO₄) Max. 20 ppm
Free chlorine (Cl) Max. 4 ppm
Residual solvents complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR