4-Nitroaniline ≥99.0% (by HPLC) indicator in non-aqueous solvents, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
p-Nitroaniline
72680-500G
BJ72680-500GEA
540.94
USD
BJ72680-500G
4-Nitroaniline ≥99.0% (by HPLC) indicator in non-aqueous solvents, Fluka™
4-Nitroaniline
Formula:
C₆H₆N₂O₂ MW: 138.13 g/mol Boiling Pt: 260 °C Melting Pt: 146-149 °C Density: 1.44 g/cm³ Flash Pt: 213 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007858 CAS Number: 100-01-6 EINECS: 202-810-1 UN: 1661 ADR: 6.1,II |
Specification Test Results
Identity (IR) | complying |
Assay (HPLC) | Min. 99.0 % |
Melting point | 146 - 150 °C |
Colour of the solution | complying |
Clearness of the solution | complying |
Residue on ignition | Max. 0.05 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...