Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Charcoal activated (from peat), Powder, Norit® SX ultra US Food chemicals codex (3rd Ed.), highly purified, steam activated and acid washed

Warning
Synonyms: Activated carbonCarbon activatedActivated charcoal

53663-100G 53663-1KG 53663-250G
BJ53663-100G BJ53663-1KG BJ53663-250G
Charcoal activated (from peat), Powder, Norit® SX ultra US Food chemicals codex (3rd Ed.), highly purified, steam activated and acid washed


Formula: C
MW: 12,01 g/mol
Melting Pt: 3550 °C
Density: 2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818
Order Now
Specification Test Results
Appearance complying
Loss on drying (110C) max. 10 %
Residue on ignition max. 8 %

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty