Ammonia solution 7 N in methanol, Fluka™

Danger

Synonyms: Ammonium hydroxide

499145-1L
BJ499145-1LEA 358.01 USD
BJ499145-1L
Ammonia solution 7 N in methanol, Fluka™
Ammonia solution
Formula: NH₃
MW: 35.05 g/mol
Melting Pt: no data available
Density: 0.779 g/cm3 (25 °C)
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
UN: 3286
ADR: 3,II

Order Now

Specification Test Results

Assay 6.5 - 7.5 mol/l
Water (Karl Fischer) Max. 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR