tert-Butyl methyl ether ≥99.0% ACS, Riedel-de Haën™

Danger

Synonyms: Methyl tert-butyl etherMethyl 1,1-dimethylethyl etherMethyl t-butyl etherEther, tert-butyl methyl2-Methyl-2-methoxypropane2-Methoxy-2-methylpropaneMTBE

443808-4X2.5L
BJ4438084X2.5LCS 412.04 USD
BJ4438084X2.5L
tert-Butyl methyl ether ≥99.0% ACS, Riedel-de Haën™
tert-Butyl methyl ether

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Beilstein Number: 1730942

Formula: (CH₃)₃COCH₃
MW: 88.15 g/mol
Boiling Pt: 55.3 °C
Melting Pt: -109 °C
Density: 0.74 g/cm3 (20 °C)
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/77/75/2297775.jpg

Order Now

Specification Test Results

Assay (GC) Min. 99.0 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.05 %
Peroxides (as H₂O₂) Max. 1 ppm
APHA Max. 10
Vapor Density 3.1 (vs air)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR