Nitric acid ≥70% ACS, Fluka™

Danger

438073-2.5L 438073-2.2L-P 438073-100ML
BJ438073-2.5LEA 179.93 USD
BJ438073-2.5L BJ438073-100ML BJ438073-2.2LP
Nitric acid ≥70% ACS, Fluka™
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 122 °C (1.013 hPa)
Melting Pt: -38 °C
Density: 1.420 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II

Order Now

Specification Test Results

Appearance (color) complying
Appearance (form) complying
Assay 68.0 - 70.0 %
Residue on ignition (SO₄) Max. 5 ppm
Arsenic (As) Max. 0.01 ppm
Iron (Fe) Max. 0.2 ppm
Heavy metals (as Pb) Max. 0.2 ppm
Chloride (Cl) Max. 0.5 ppm
Sulfate (SO₄) Max. 1 ppm
APHA Max. 10
Ignition Residue ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR