Zinc acetate dihydrate ≥98% ACS, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Zinc diacetate dihydrate
, Acetic acid zinc salt dihydrate
, Zinc (II) acetate dihydrate
383058-25KG
BJ383058-25KGEA
2503.7
USD
BJ383058-25KG
Zinc acetate dihydrate ≥98% ACS, Fluka™
Zinc acetate dihydrate
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Beilstein Number: 3732513
RTECS Number: 209-170-2
UNSPSC Code: ZG8750000
Formula:
(H₃CCOO)₂Zn·2H₂O MW: 219.51 g/mol Melting Pt: 100 °C Density: 1.750 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00066961 CAS Number: 5970-45-6 UN: 3077 ADR: 9,III |
Specification Test Results
Identity (IR) | complying |
Assay | 98.0 - 101.0 % |
pH (5 %, 25 °C) | 6.0 - 7.0 |
in CH₃COOH insoluble matters | Max. 0.005 % |
Calcium (Ca) | Max. 0.005 % |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | Max. 0.01 % |
Magnesium (Mg) | Max. 0.005 % |
Sodium (Na) | Max. 0.05 % |
Lead (Pb) | Max. 0.002 % |
Chloride (Cl) | Max. 5 ppm |
Sulfate (SO₄) | Max. 0.005 % |
Appearance (color) | complying |
Appearance (form) | complying |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...