1-Butanol ≥99.4% ACS, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Butyl alcohol
, Methylolpropane
, Butyl hydroxide
, n-Butanol
, n-Butyl alcohol
, Propylcarbinol
, 1-Hydroxybutane
, 1-Hydroxy butane
, Propyl carbinol
, Butan-1-ol
, 1-Butyl alcohol
, Butyl Alcohol
, n-Butyl Alcohol
, n-Butan-1-ol
360465-2.5L
360465-4X2.5L
BJ360465-2.5LEA
217.01
USD
BJ360465-2.5L
BJ3604654X2.5L
1-Butanol ≥99.4% ACS, Riedel-de Haën™
1-Butanol
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Beilstein Number: 969148
RTECS Number: EO1400000
UNSPSC Code: 12191601
Formula:
H₃C(CH₂)₂CH₂OH MW: 74.12 g/mol Boiling Pt: 119 °C (1.013 hPa) Melting Pt: <-90 °C Density: 0.81 g/cm³ (20 °C) Flash Pt: 9.4 %(V) Storage Temperature: Ambient |
MDL Number:
MFCD00002964 CAS Number: 71-36-3 EINECS: 200-751-6 UN: 1120 ADR: 3,III |
Specification Test Results
Autoignition Temperature | 649 °F |
Identity (IR) | complying |
Assay (GC) | Min. 99.40 % |
Non-volatile matter | Max. 0.005 % |
Water (Karl Fischer) | Max. 0.1 % |
Titrable Acid | Max. 0.0008 meq/g |
Di-n-butyl ether (GC) | Max. 0.2 % |
n-Butyraldehyde (GC) | Max. 0.01 % |
APHA | Max. 10 |
Explosion Limit | 11.2 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...