Rinsing solution I, CHROMASOLV™, LabReady™, Solvent Blend rinsing agent for LC-MS, Riedel-de Haën™

Danger

34689-1L
BJ34689-1LEA 100.13 USD
BJ34689-1L
Rinsing solution I, CHROMASOLV™, LabReady™, Solvent Blend rinsing agent for LC-MS, Riedel-de Haën™
Rinsing solution I

  • High purity chromatography solvents
  • Suitability tested for specified use from HPLC up to UHPLC-MS applications.

Flash Pt: 22 °C
Storage Temperature: Ambient
UN: 1219
ADR: 3,II

Order Now

Specification Test Results

Assay of water 49 - 51 Vol.%
Assay of water 49 - 51 Vol.%
Identity 2-propanol (GC) complying
Identity 2-propanol (GC) complying
Non-volatile matter Max. 0.001 %
Silver (Ag) Max. 0.1 ppm
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Calcium (Ca) Max. 0.1 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Sodium (Na) Max. 0.1 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Tin (Sn) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Organic impurities GC Max. 0.1 Fl.%
Absobance at 254 nm Max. 0.008
Absorbance at 230 nm Max. 0.045
Absorbance at 210 nm Max. 0.096
Suitability for the LC-MS complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR