1,2-Dimethoxyethane ≥99.9% inhibitor-free, CHROMASOLV™ for HPLC, Riedel-de Haën™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Dimethyl glycol
307432-1L
BJ307432-1LEA
161.87
USD
BJ307432-1L
1,2-Dimethoxyethane ≥99.9% inhibitor-free, CHROMASOLV™ for HPLC, Riedel-de Haën™
1,2-Dimethoxyethane
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Formula:
C₄H₁₀O₂ MW: 90.12 g/mol Boiling Pt: 85 °C (1.013 hPa) Melting Pt: -58 °C Density: 0.87 g/cm³ (20 °C) Flash Pt: -2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008502 CAS Number: 110-71-4 EINECS: 203-794-9 UN: 2252 ADR: 3,II |
Specification Test Results
Identity (IR) | complying |
Assay (GC) | Min. 99.85 % |
Non-volatile matter | Max. 0.0005 % |
Water (Karl Fischer) | Max. 0.03 % |
Peroxides (as H₂O₂) | Max. 15 ppm |
Absorbance at 400 nm | Max. 0.01 |
Absorbance at 350 nm | Max. 0.01 |
Absorbance at 300 nm | Max. 0.03 |
Absorbance at 250 nm | Max. 0.20 |
Absorbance at 220 nm | Max. 1.0 |
Vapor Density | 3.1 (20 °C, vs air) |
Refractive Index | n20/D 1.379 (lit.) |
Explosion Limit | 10.4 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...