Cyclopentane ≥75% (Cyclopentane basis), CHROMASOLV™ for HPLC, Riedel-de Haën™

Danger

Synonyms: Pentamethylene

270601-1L
BJ270601-1LEA 438.85 USD
BJ270601-1L
Cyclopentane ≥75% (Cyclopentane basis), CHROMASOLV™ for HPLC, Riedel-de Haën™
Cyclopentane

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formula: C₅H₁₀
MW: 70.13 g/mol
Melting Pt: -94 °C
Density: 0.751 g/cm3 (25 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00001356
CAS Number: 287-92-3
EINECS: 206-016-6
UN: 1146
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/27/15/772715.jpg

Order Now

Specification Test Results

Autoignition Temperature 682 °F
Identity (IR) complying
Assay (GC) Min. 75.0 %
Non-volatile matter Max. 0.0003 %
Water (Karl Fischer) Max. 0.02 %
Absorbance at 400 nm Max. 0.01
Absorbance at 240 nm Max. 0.01
Absorbance at 220 nm Max. 0.10
Absorbance at 210 nm Max. 0.50
Absorbance at 198 nm Max. 1.0
Vapor Pressure 5.32 psi ( 20 °C)
Explosion Limit 8.7 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR