Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™

Danger

Synonyms: Ethylene glycol monomethyl ether

270482-1L
BJ270482-1LEA 229.65 USD
BJ270482-1L
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99.9%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Ethylene glycol monomethyl ether (2-Methoxyethanol)

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formula: C₃H₈O₂
MW: 76.1 g/mol
Boiling Pt: 124 °C
Melting Pt: -85 °C
Density: 0.97 g/cm3 (20 °C)
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00002867
CAS Number: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III

Order Now

Specification Test Results

Vapor Density 2.62 (vs air)
Identity (IR) complying
Assay (GC) Min. 99.90 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.1 %
Absorbance at 400 nm Max. 0.01
Absorbance at 300 nm Max. 0.01
Absorbance at 260 nm Max. 0.05
Absorbance at 240 nm Max. 0.20
Absorbance at 213 nm Max. 1.0
Explosion Limit 24.5 %
Refractive Index n20/D 1.402 (lit.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR