Cadmium(II) chloride hemipentahydrate 79.5-81.0% ACS, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
239208-500G
BJ239208-500GEA
334.88
USD
BJ239208-500G
Cadmium(II) chloride hemipentahydrate 79.5-81.0% ACS, Fluka™
Cadmium (II) chloride hemipentahydrate
Formula:
CdCl₂·2,5H₂O MW: 228.35 g/mol Density: 3.327 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00799071 CAS Number: 7790-78-5 UN: 2570 ADR: 6.1,III |
Specification Test Results
Vapor Pressure | 10 mmHg ( 656 °C) |
Assay of CdCl₂ | 79.5 - 81.0 % |
Water insoluble matter | Max. 0.005 % |
Calcium (Ca) | Max. 0.005 % |
Copper (Cu) | Max. 5 ppm |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | Max. 0.02 % |
Sodium (Na) | Max. 0.05 % |
Lead (Pb) | Max. 0.005 % |
Zinc (Zn) | Max. 0.05 % |
Ammonium (NH₄) | Max. 0.005 % |
Nitrate, nitrite (as NO₃) | Max. 0.003 % |
Sulfate (SO₄) | Max. 0.005 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...