Mercury(I) chloride ≥99.5% ACS, Fluka™
Supplier: Honeywell Research Chemicals
|
Warning
|
Synonyms:
Dimercury dichloride
230405-100G
BJ230405-100GEA
205.73
USD
BJ230405-100G
Mercury(I) chloride ≥99.5% ACS, Fluka™
Mercury (I) chloride
Formula:
Cl₂Hg₂ MW: 472.09 g/mol Melting Pt: 400 °C (Sublimation point) Density: 7.15 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011043 CAS Number: 10112-91-1 EINECS: 233-307-5 UN: 2025 ADR: 6.1,III |
Specification Test Results
Assay | min. 99.5 % |
Assay | Min. 99.5 % |
Residue on ignition a. reducing with HCOOH (SO₄)* | Max. 0.02 % |
Mercury(II) (as HgCl₂) | Max. 0.01 % |
Sulfate (SO₄) | Max. 0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...