Cadmium (II) sulfate 8/3-hydrate ≥99.0% (by complexometric analysis), puriss. p.a. ACS, Fluka™

Supplier: Honeywell Research Chemicals

Danger

Synonyms: Cadmium sulphate, 8/3-hydrateCadmium sulfate, 8/3-hydrateCadmium sulphate octahydrateCadmium sulfate octahydrate

20920-500G 20920-100G 20920-25G
BJ20920-500GEA 237.72 USD
BJ20920-500G BJ20920-100G BJ20920-25G
Cadmium (II) sulfate 8/3-hydrate ≥99.0% (by complexometric analysis), puriss. p.a. ACS, Fluka™
Cadmium (II) sulphate 8/3-hydrate
Formula: 3CdSO₄·8H₂O
MW: 256.51 g/mol
Boiling Pt: Dec.
Melting Pt: 41,5 °C
Density: 3,09 g/cm3
Storage Temperature: Ambient
MDL Number: MFCD00151655
CAS Number: 7790-84-3
EINECS: 233-331-6
UN: 2570
ADR: 6.1,II
Merck Index: 13,01627

Order Now

Specification Test Results

Assay (complexometric) 99.0 - 102.0 %
Water insoluble matter max. 0.005 %
Arsenic (As) max. 2 ppm
Calcium (Ca) max. 50 ppm
Cobalt (Co) max. 5 ppm
Chromium (Cr) max. 5 ppm
Copper (Cu) max. 5 ppm
Iron (Fe) max. 5 ppm
Potassium (K) max. 50 ppm
Magnesium (Mg) max. 5 ppm
Manganese (Mn) max. 5 ppm
Sodium (Na) max. 50 ppm
Nickel (Ni) max. 10 ppm
Lead (Pb) max. 20 ppm
Zinc (Zn) max. 50 ppm
Chloride (Cl) max. 10 ppm
Total nitrogen (as NO₃) max. 30 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR