Hexachloroplatinic(IV) acid hexahydrate ≥37.50% (Pt basis) ACS, Fluka™
Supplier: Honeywell Research Chemicals
|
Danger
|
Synonyms:
Dihydrogen hexachloroplatinate (IV) hexahydrate
206083-5G
206083-25G
206083-1G
BJ206083-25GEA
2408.89
USD
BJ206083-25G
BJ206083-1G
BJ206083-5G
Hexachloroplatinic(IV) acid hexahydrate ≥37.50% (Pt basis) ACS, Fluka™
Hexachloroplatinic (IV) acid hexahydrate
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Formula:
H₂PtCl₆·6H₂O MW: 517.9 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00149910 CAS Number: 18497-13-7 EINECS: 241-010-7 UN: 2507 ADR: 8,III |
Specification Test Results
Appearance | complying |
Platinum (Pt) | Min. 37.50 % |
Solubility in C₂H₅OH | complying |
Alkalis (as Sulfates) | Max. 0.05 % |
Application test | complying |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...