2,2,2-Trifluoroethanol eluent additive, for LC-MS, Fluka™

Danger

Synonyms: TFE

05841-50ML
BJ05841-50MLEA 249.7 USD
BJ05841-50ML
2,2,2-Trifluoroethanol eluent additive, for LC-MS, Fluka™
2,2,2-Trifluoroethanol
Formula: C₂H₃F₃O
MW: 100.04 g/mol
Boiling Pt: 73 - 77 °C
Melting Pt: -44 °C
Density: 1.383 g/cm3 (25 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00004672
CAS Number: 75-89-8
EINECS: 200-913-6
UN: 1986
ADR: 3 (6.1),III

Order Now

Specification Test Results

Identity (NMR) complying
Assay (GC) Min. 99.8 %
Density (D 20/4) 1.390 - 1.392
Suitability for the LC-MS complying
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 1 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
Explosion Limit 42 %
Refractive Index n20/D 1.3 (lit.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR