Bromocresol green solution indicator, Ready-to-Use, Fluka™

Supplier: Honeywell Research Chemicals

Danger

02559-500ML
BJ02559-500MLEA 132.87 USD InStock
BJ02559-500ML
Bromocresol green solution indicator, Ready-to-Use, Fluka™
Bromocresol green
Formula: C₂₁H₁₄Br₄O₅S
MW: 698.02 g/mol
Melting Pt: 89,5 °C
Density: 0,79 g/cm3 (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00005874
CAS Number: 76-60-8
EINECS: 200-661-7
UN: 1219
ADR: 3,II
Merck Index: 13,01370
https://us.vwr.com/stibo/web/std.lang.all/13/36/2301336.jpg

Order Now

Specification Test Results

Appearance of the substance complying
Density (D 20/4) 0.775 - 0.804
Absorption maximum 408 - 438 nm
Absorbtion (1%/1cm) max. 0.50

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR