[Lys(Ac)5]-Histone H4 (1-21)-GGK,Biotin

Supplier: AnaSpec
AS-64844-1
103006-540EA 482.31 USD
103006-540
[Lys(Ac)5]-Histone H4 (1-21)-GGK,Biotin
Proteins and Peptides
This peptide is histone H4 amino acids 1 to 21, acetylated at Lys-5 and at the N-terminus. This peptide also contains a C-terminal GG linker, followed by a biotinylated lysine. The acetylation of histone H4 causes structural changes that play a crucial role in amplifying the binding of transcription factors to specific recognition sites within the nucleosome.
Sequence:Ac-SGRG-K(Ac)-GGKGLGKGGAKRHRKV-GGK(Biotin)
MW:2644.1 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR