Histone H3 (69-89)-K, Biotin

Supplier: AnaSpec
AS-65297-1
103009-380EA 403.38 USD
103009-380
Histone H3 (69-89)-K, Biotin
Proteins and Peptides
This peptide is histone H3 (69-89) biotinylated through the epsilon side chain of a C-terminal Lys. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:RLVREIAQDFKTDLRFQSSAV-K(Biotin)
MW:2834.3 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR