[Lys(Ac)36]-Histone H3 (21-44)-GK,Biotin

Supplier: AnaSpec
AS-64936-1
103008-596EA 599.23 USD
103008-596
[Lys(Ac)36]-Histone H3 (21-44)-GK,Biotin
Proteins and Peptides
This peptide is Histone H3 (21-44). It is acetylated at lysine 36 with a C-terminal G linker followed by a biotinylated lysine. The acetylation of histone H3 at lysine (K) 36 is highly conserved in mammals and is mainly localized to the promoters of RNA polymerase II-transcribed genes. The transition between the acetylation and methylation of K36 acts as an “acetyl/methyl switch”, controlling chromatin function along transcription units. Provided at >95% peptide purity, this peptide was dissolved in distilled water at 1 mg/ml and re-lyophilized to powder form.
Sequence:ATKAARKSAPATGGV-K(Ac)-KPHRYRPG-GK(Biotin)
MW:2959.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR