[Lys(Me2)9]-Histone H3 (3-17)

Supplier: AnaSpec
AS-64373-1
103008-024EA 314.23 USD
103008-024
[Lys(Me2)9]-Histone H3 (3-17)
Proteins and Peptides
This peptide is Histone H3 amino acid residues 1 to 10 di-methylated at Lys-9.
Sequence:TKQTAR-K(Me2)-STGGKAPR
MW:1614.8 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR