Histone H3 (1-21)

Supplier: AnaSpec
AS-61701
103006-910EA 314.23 USD
103006-910
Histone H3 (1-21)
Proteins and Peptides
This peptide is derived from Histone H3 1-21 amino acids, and is usually used as a substrate for methylation and acetylation assays.
Sequence:ARTKQTARKSTGGKAPRKQLA
MW:2254.6 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR