TP53 Q9NP68, p53 Mutant Form (372-389), Lys382 (Ac), Anaspec Inc.

Supplier: Anaspec Inc
AS-62121
103007-198EA 206.08 USD
103007-198
TP53 Q9NP68, p53 Mutant Form (372-389), Lys382 (Ac), Anaspec Inc.
Proteins and Peptides
This peptide is derived from the tumor suppressor p53 mutant with the acetylated Lys on the side chain.
Sequence:KKGQSTSRHK-K(Ac)-LMFKTEG
MW:2133.5 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR