Bid BH3 Peptide, FAM (Carboxyfluorescein)

Supplier: AnaSpec
AS-61712
103006-924EA 543.69 USD
103006-924
Bid BH3 Peptide, FAM (Carboxyfluorescein)
Proteins and Peptides
This is a 5-FAM-labeled Bid BH3 peptide. Bid is a pro-apoptotic member of the 'BH3-only' subset of the BCL-2 family proteins that constitute a critical control point in apoptosis. Bid interconnects extrinsic pathway TNFR1 and Fas death signals to the mitochondrial amplification of the intrinsic pathway. The references listed below belong to the unlabeled Bid BH3.
Sequence:5-FAM-EDIIRNIARHLAQVGDSMDR
MW:2667.9 Da
% peak area by HPLC:95
Storage condition:-20° C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR