LiChroCART® Stainless Steel Purospher® RP-18 HC Cartridges, MilliporeSigma
Supplier: MilliporeSigma
LiChroCART® Purospher®
1.51436.0001 1.51436.0001
10143-956EA 779.1 USD
CA1.51436.0001 10143-956
LiChroCART® Stainless Steel Purospher® RP-18 HC Cartridges, MilliporeSigma
Chromatography Columns
Purospher® RP-18 HC is excellently suitable for high resolution separation of explosives and related compounds.
The determination of explosives is of great importance, including the quantification of their by products like nitrotoluene and nitrophenol, as well as nitroaminotoluene and ami-notoluene. Purospher® RP-18 HC is also suitable for the separation of picric acid from hexyl and ethyleneglycol dinitrate from ethyleneglycol nitrate.