Tissue-Tek® Quick Ray™ Tissue Microarray System, Electron Microscopy Sciences
62595-09 62595-06 62595-04 62595-07 62595-02 62595-11 62595-10 62595-03 62595-05 62595-08
103303-246EA 94.76 USD
103303-246 103301-978 103303-248 103306-316 103306-318 103306-312 103306-314 103303-250 103306-264 103303-252
Tissue-Tek® Quick Ray™ Tissue Microarray System, Electron Microscopy Sciences
The Tissue Microarray are a collection of multiple tissue cores that are arranged in an XY fashion inside of a paraffin block allowing for histological analysis. The construction of a TMA block can be done on any clean counter, and does not require a dedicated space.
  • Compact design and versatility
  • Ready-to-use paraffin recipient block
The Tissue-Tek® Quick-Ray™ technique produces superior quality TMAs in less time than with traditional methods, and accomplishes finished products at a fraction of the cost
The finished block is sectioned using a microtome; the sections are mounted on glass slides and then stained. Each TMA block can be cut into 300 – 400 sections at 4 – 5 µm, which can then be subjected to independent tests. Common tests include immunohistochemistry and fluorescent in situ hybridization.
The Tissue-Tek® Quick-Ray™ uses a hollowed tip to remove tissue cores as small as 1 mm from paraffin-embedded tissue (donor block). The extracted tissue is then inserted into a preformed recipient block–the Quick-Ray™ System is the only system available that uses a preformed paraffin recipient block.
Delivery information: Includes 1 puncher, 4 paraffin recipient blocks (1,2,3 and 5 mm), 1 basemold, 4 punch tips (1,2,3 and 5 mm) and 1 punch guide (1 mm).