    Sort by:
  • Search Within Results
Atazanavir
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  BioVision
Description:   Atazanavir is a potent inhibitor of HIV protease (Ki of 2.66 nM). Atazanavir binds to HIV-1 protease preventing the cleavage of gag and gag-pol polypr...