    Sort by:
  • Search Within Results
Isothiazole
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   Isothiazole, 97%, CAS No 288-16-4, Molecular weight 85.13, Molecular formula C3 H3 N S, Appearance (Color) Clear colorless to yellow, Appearance (Form) Liquid, Infrared spectrum Authentic, GC >=96.0 %, Synonyms: 1,2-Thiazole, Size: 5G