    Sort by:
  • Search Within Results
4-Azaphthalide
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   4-Azaphthalide, Purity: 98%, CAS Number: 5657-51-2, Molecular Formula: C7H5NO2, Formula Weight: 135.12g/mol, Synonyms: 2(Hydroxymethyl)nicotinic acid gamma-lactone, Furo[3,4-b]pyridin-5(7H)-one, Physical Form: Powder or crystalline powder, Color: White to light brown, Size: 5g