    Sort by:
  • Search Within Results
(R)-3-Phenylmorpholine hydrochloride
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   (R)-3-Phenylmorpholine hydrochloride, Purity: 97%, CAS Number: 1624261-27-3, Molecular Formula: C10H13NO.ClH, Formula Weight: 199.68g/mol, Physical Form: Powder, Color: White to yellow, Size: 5g