    Sort by:
  • Search Within Results
1,4-Cineole
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   1,4-Cineole, 85%, CAS No 470-67-7, MW 154.25, Molecular formula C10 H18 O, Color Clear colorless, Form Liquid, Infrared spectrum Authentic, Refractive index 1.4450 to 1.4560 (20 deg C, 589 nm), GC >=80 %, Synonyms: 1-Isopropyl-4-methyl-7-oxabicyclo[2.2.1]heptane, Size: 100g