Nylon Cloth Labels for Vial Tops, Round , LABXPERT™ B-499, GA International
Supplier: GA International
LABXPERT™
60950
80090-390EA 292.74 USD
80090-390
Nylon Cloth Labels for Vial Tops, Round , LABXPERT™ B-499, GA International
Labels
LABXPERT™ nylon cloth printable labels is used for laboratory identification and the material is UL recognized with its respective ribbon.
  • Good moisture resistance
  • Permanent acrylic
  • Excellent water and oil resistance
These labels withstands extremely low temperatures and has high adhesion capacity.