    Sort by:
  • Search Within Results
Millon's reagent, for the detection of Tyrosine
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Acros Organics
Description:   Millon's reagent, for the detection of Tyrosine, CAS No 10045-94-0, MW 324.60, Molecular formula Hg N2 O6, Appearance (Color) Clear colorless to light yellow, Form Solution, Sensitivity passes test, Density 1.2 to 1.5 kg/l, Size: 100ml