    Sort by:
  • Search Within Results
Magnesium matrix modifier
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  Honeywell Research Chemicals
Description:   Magnesium matrix modifier solution, Grade: Analytical, Density: 1.093 g/cm3, Synonyms: Mixture, Spectroscopy Reagents, 2% Mg in 5 wt. % HNO3, 99.9995%, Size: 100ML