Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Proto® 440SS Tool Cabinets 12 & 15 Drawer Models, Stanley® Products

Supplier: ORS NASCO, INC.
5011150686 5011150693
Smooth, sleek, and durable red paint finish

  • Maximum weight capacity for each drawer is 100 lbs
  • Aluminum extruded drawer pulls
  • Recessed drawer pulls and 0.040" drawer front and body
  • Heavy-duty key
  • Four-way channel bottom panel
  • Polyurethane casters: 5" Dia. x 2" W
  • Heavy-duty, easy grip handle

Drawer dimensions for the 12 Drawer model are: (1) 11 1/8W x 16 1/8D x 315/16"H; (4) 5W x 16 1/8D x 1 7/8"H; (2) 23 1/4W x 16 1/8D x 1 7/8" H; (4) 23 1/4W x 16 1/8D x 3 15/16"H; (1) 23 1/4W x 16 1/8D x 8 1/16"H.
Drawer dimensions for the 15 Drawer model are: (1) 11 1/8W x 16 1/8D x 8 1/16"H; (1) 23 1/4W x 6 1/8D x 8 1/16"H; (1) 37 3/4W x 16 1/8D x 3 15/16"H; (2) 11 1/8W x 16 1/8D x 3 15/16"H; (2) 23 1/4W x 16 1/8D x 3 15/16"H; (4) 11 1/8W x 16 1/8D x 1 7/8"H; (4) 23 1/4W x 16 1/8D x 1 7/8"H.

All cabinets include a vinyl top, drawer liners, and ball bearing type slides.



 

ORDER

Expand All / Collapse All
Material Capacity Depth Height Width Color No. of Drawers Weight Capacity (Volume) Supplier No. VWR Catalog Number Unit Price Quantity
Steel 100 lbs./Drawer; 450 lbs./Caster 18" 42" 41" Red 15 290.5 lbs. 14,362 cu. in. 5011150693 300011-362 Each Retrieving
Steel 100 lbs./Drawer; 450 lbs./Caster 18" 42" 27" Red 12 188 lbs. 9,420 cu. in. 5011150686 300011-424 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty