Leica CM3050 S ensures efficient specimen processing by offering fast and reliable sectioning results
- Convenient section thickness selection from outside the cryochamber
- Indication of total section thickness
- Reproducible high-quality thin sections via step motor specimen feed
- Programmable reverse section counter
- Spacious cryochamber, easy to clean and disinfect
- Low-maintenance design due to convenient access to the cooling system from outside the cryostat housing
- Low-maintenance, durable refrigerating system
- Encapsulated microtome to support efficient spray disinfection
Max specimen size: 40 x 55 mm
An innovative heat insulation system guarantees constant stable temperatures and less power consumption, therefore helping to keep running costs low. With its versatility and easy operation the Leica CM3050 S even meets the highest demands perfectly.The Leica CM3050 S cryostat is equipped with sectioning motor and available with and without object cooling.